Imprimer

Anti-IKBKG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 55AT986.5.78]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031047-APCEA 720 EUR
USBI031047-APC
Anti-IKBKG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 55AT986.5.78]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: Monoclonal
Clone: 55AT986.5.78
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR