Imprimer

Anti-INA Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 257CT7.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031054-APEA 711 EUR
USBI031054-AP
Anti-INA Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 257CT7.1.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50

Type: Primary
Antigen: INA (internexin neuronal intermediate filament protein, alpha)
Clonality: Monoclonal
Clone: 257CT7.1.2
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR