Imprimer

Anti-Interferon alpha Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 278CT1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031057-HRPEA 711 EUR
USBI031057-HRP
Anti-Interferon alpha Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 278CT1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: Interferon alpha
Clonality: Monoclonal
Clone: 278CT1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR