Imprimer

Anti-ITGAX/CD11c Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 428CT12.2.6.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031061-HRPEA 711 EUR
USBI031061-HRP
Anti-ITGAX/CD11c Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 428CT12.2.6.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ITGAX/CD11c
Clonality: Monoclonal
Clone: 428CT12.2.6.4
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR