Imprimer

Anti-Kappa light chain Mouse Monoclonal Antibody [clone: HP6053]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031066EA 495 EUR
USBI031066
Anti-Kappa light chain Mouse Monoclonal Antibody [clone: HP6053]
Anticorps
Recommended dilution: Western Blot: 1:100-1:1000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: Kappa light chain
Clonality: Monoclonal
Clone: HP6053
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR