Imprimer

Anti-Kappa light chain Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HP6053]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031066-HRPEA 711 EUR
USBI031066-HRP
Anti-Kappa light chain Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HP6053]
Anticorps
Type: Primary
Antigen: Kappa light chain
Clonality: Monoclonal
Clone: HP6053
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR