Imprimer

Anti-KIT Mouse Monoclonal Antibody [clone: 566CT8.5.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031070EA 587 EUR
USBI031070
Anti-KIT Mouse Monoclonal Antibody [clone: 566CT8.5.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-16000

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone: 566CT8.5.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR