Imprimer

Anti-Lambda light chain Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HP6054]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031075-APCEA 711 EUR
USBI031075-APC
Anti-Lambda light chain Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HP6054]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:500-16000; Immunochemistry: 1:10-50

Type: Primary
Antigen: Lambda light chain
Clonality: Monoclonal
Clone: HP6054
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR