Imprimer

Anti-LIN28A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 55CT58.12.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031089-APCEA 711 EUR
USBI031089-APC
Anti-LIN28A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 55CT58.12.1]
Anticorps
Recommended dilution: FLISA: 1:10000; Western Blot: 1:200-2000; Immunochemistry: 1:50-200

Type: Primary
Antigen: LIN28A (lin-28 homolog A (C. elegans))
Clonality: Monoclonal
Clone: 55CT58.12.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR