Imprimer

Anti-LOX Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 624CT23.7.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031090-FITCEA 711 EUR
USBI031090-FITC
Anti-LOX Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 624CT23.7.3]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: Monoclonal
Clone: 624CT23.7.3
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR