Imprimer

Anti-LTF Mouse Monoclonal Antibody (Biotin) [clone: 119CT80.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031095-BIOTINEA 720 EUR
USBI031095-BIOTIN
Anti-LTF Mouse Monoclonal Antibody (Biotin) [clone: 119CT80.1.1]
Anticorps
Recommended dilution: ELISA: 1:3000; Western Blot: 1:1000-16000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: Monoclonal
Clone: 119CT80.1.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR