Imprimer

Anti-MAP Kinase 15 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 168CT10.6.6]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031100-APCEA 711 EUR
USBI031100-APC
Anti-MAP Kinase 15 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 168CT10.6.6]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MAP Kinase 15
Clonality: Monoclonal
Clone: 168CT10.6.6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR