Imprimer

Anti-MAP2 Mouse Monoclonal Antibody [clone: 159CT34.12.3.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031102EA 487 EUR
USBI031102
Anti-MAP2 Mouse Monoclonal Antibody [clone: 159CT34.12.3.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:450

Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality: Monoclonal
Clone: 159CT34.12.3.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR