Imprimer

Anti-MAPK3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 327CT18.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031110-APCEA 720 EUR
USBI031110-APC
Anti-MAPK3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 327CT18.1.2]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:120-1000

Type: Primary
Antigen: MAPK3 (mitogen-activated protein kinase 3)
Clonality: Monoclonal
Clone: 327CT18.1.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR