Imprimer

Anti-MLANA Mouse Monoclonal Antibody [clone: 610CT14.6.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031111EA 587 EUR
USBI031111
Anti-MLANA Mouse Monoclonal Antibody [clone: 610CT14.6.4]
Anticorps
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: MLANA (melan-A)
Clonality: Monoclonal
Clone: 610CT14.6.4
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR