Imprimer

Anti-MASTL Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 235CT7.8.2.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031113-APEA 852 EUR
USBI031113-AP
Anti-MASTL Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 235CT7.8.2.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MASTL (microtubule associated serine/threonine kinase-like)
Clonality: Monoclonal
Clone: 235CT7.8.2.3
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR