Imprimer

Anti-MET Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 6AT44.279]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031119-FITCEA 562 EUR
USBI031119-FITC
Anti-MET Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 6AT44.279]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MET (met proto-oncogene)
Clonality: Monoclonal
Clone: 6AT44.279
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR