Imprimer

Anti-MMP14 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 133CT15.10.5.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031127-FITCEA 619 EUR
USBI031127-FITC
Anti-MMP14 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 133CT15.10.5.1]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: MMP14 (matrix metallopeptidase 14 (membrane-inserted))
Clonality: Monoclonal
Clone: 133CT15.10.5.1
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR