Imprimer

Anti-Mobc Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: ID:19CT.2.8.14]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031133-APEA 616 EUR
USBI031133-AP
Anti-Mobc Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: ID:19CT.2.8.14]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: Mobc
Clonality: Monoclonal
Clone: ID:19CT.2.8.14
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR