Imprimer

Anti-Mouse Bmi1 Mouse Monoclonal Antibody [clone: 353CT22.4.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031134EA 445 EUR
USBI031134
Anti-Mouse Bmi1 Mouse Monoclonal Antibody [clone: 353CT22.4.4]
Anticorps
Recommended dilution: Western Blot: 1:200-1600

Type: Primary
Antigen: Mouse Bmi1
Clonality: Monoclonal
Clone: 353CT22.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR