Imprimer

Anti-Myc Tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031143-APCEA 728 EUR
USBI031143-APC
Anti-Myc Tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 9E10]
Anticorps
Recommended dilution: FLISA: 1:3000; Western Blot: 1:2000

Type: Primary
Antigen: Myc Tag (Myc tag)
Clonality: Monoclonal
Clone: 90000000000
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR