Imprimer

Anti-MYOG Mouse Monoclonal Antibody (Biotin) [clone: 304CT11.6.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031148-BIOTINEA 711 EUR
USBI031148-BIOTIN
Anti-MYOG Mouse Monoclonal Antibody (Biotin) [clone: 304CT11.6.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: MYOG (myogenin (myogenic factor 4))
Clonality: Monoclonal
Clone: 304CT11.6.1
Conjugation: Biotin
Epitope: N-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR