Imprimer

Anti-NPHS1 Mouse Monoclonal Antibody [clone: 174CT2.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031156EA 527 EUR
USBI031156
Anti-NPHS1 Mouse Monoclonal Antibody [clone: 174CT2.1.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-200; Immunofluorescence: 1:10-50

Type: Primary
Antigen: NPHS1 (nephrosis 1, congenital, Finnish type (nephrin))
Clonality: Monoclonal
Clone: 174CT2.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR