Imprimer

Anti-NLRP12 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 228CT4.1.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031160-APEA 619 EUR
USBI031160-AP
Anti-NLRP12 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 228CT4.1.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1500

Type: Primary
Antigen: NLRP12 (NLR family, pyrin domain containing 12)
Clonality: Monoclonal
Clone: 228CT4.1.3
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR