Imprimer
Anti-NOTCH3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 487CT6.9.2]
Fournisseur: United States Biological
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBI031162-FITCEA 852 EUR
USBI031162-FITC
Anti-NOTCH3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 487CT6.9.2]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Type: Primary
Antigen: NOTCH3 (notch 3)
Clonality: Monoclonal
Clone: 487CT6.9.2
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human