Imprimer

Anti-NOTCH3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 487CT6.9.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031162-PEEA 711 EUR
USBI031162-PE
Anti-NOTCH3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 487CT6.9.2]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250
Do not freeze! Light sensitive

Type: Primary
Antigen: NOTCH3 (notch 3)
Clonality: Monoclonal
Clone: 487CT6.9.2
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR