Imprimer

Anti-NPM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3A9F1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031163-APCEA 562 EUR
USBI031163-APC
Anti-NPM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3A9F1]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:460

Type: Primary
Antigen: NPM1 (nucleophosmin (nucleolar phosphoprotein B23, numatrin))
Clonality: Monoclonal
Clone: 3A9F1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR