Imprimer

Anti-NTRK1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 185CT47.6.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031166-HRPEA 711 EUR
USBI031166-HRP
Anti-NTRK1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 185CT47.6.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NTRK1 (neurotrophic tyrosine kinase, receptor, type 1)
Clonality: Monoclonal
Clone: 185CT47.6.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR