Imprimer

Anti-NUAK2 Mouse Monoclonal Antibody [clone: 378CT15.4.5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031167EA 527 EUR
USBI031167
Anti-NUAK2 Mouse Monoclonal Antibody [clone: 378CT15.4.5]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NUAK2 (NUAK family, SNF1-like kinase, 2)
Clonality: Monoclonal
Clone: 378CT15.4.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR