Imprimer
Anti-NUP50 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 247CT10.2.5]
Fournisseur: United States Biological
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBI031170-FITCEA 616 EUR
USBI031170-FITC
Anti-NUP50 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 247CT10.2.5]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:1560
Type: Primary
Antigen: NUP50 (nucleoporin 50kDa)
Clonality: Monoclonal
Clone: 247CT10.2.5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human