Imprimer
Anti-LRRK2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 133AT720]
Fournisseur: United States Biological
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBI031177-PEEA 852 EUR
USBI031177-PE
Anti-LRRK2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 133AT720]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze! Light sensitive
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: Monoclonal
Clone: 133AT720
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse