Imprimer

Anti-PDGFRB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 275CT1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031183-PEEA 711 EUR
USBI031183-PE
Anti-PDGFRB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 275CT1]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:830
Do not freeze! Light sensitive

Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: Monoclonal
Clone: 275CT1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR