Imprimer

Anti-PDK2 Mouse Monoclonal Antibody (Biotin) [clone: 180CT10.2.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031185-BIOTINEA 720 EUR
USBI031185-BIOTIN
Anti-PDK2 Mouse Monoclonal Antibody (Biotin) [clone: 180CT10.2.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PDK2 (pyruvate dehydrogenase kinase, isozyme 2)
Clonality: Monoclonal
Clone: 180CT10.2.3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR