Imprimer

Anti-PINK1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 38CT20.8.5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031196-FITCEA 720 EUR
USBI031196-FITC
Anti-PINK1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 38CT20.8.5]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: Monoclonal
Clone: 38CT20.8.5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR