Imprimer

Anti-PSMA5 Mouse Monoclonal Antibody (Biotin) [clone: 426CT8.5.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031210-BIOTINEA 720 EUR
USBI031210-BIOTIN
Anti-PSMA5 Mouse Monoclonal Antibody (Biotin) [clone: 426CT8.5.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PSMA5 (proteasome (prosome, macropain) subunit, alpha type, 5)
Clonality: Monoclonal
Clone: 426CT8.5.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR