Imprimer

Anti-RPS6KA1 Mouse Monoclonal Antibody (Biotin) [clone: 205CT18.3.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031222-BIOTINEA 711 EUR
USBI031222-BIOTIN
Anti-RPS6KA1 Mouse Monoclonal Antibody (Biotin) [clone: 205CT18.3.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: RPS6KA1 (ribosomal protein S6 kinase, 90kDa, polypeptide 1)
Clonality: Monoclonal
Clone: 205CT18.3.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR