Imprimer

Anti-RPS6KA3 Mouse Monoclonal Antibody [clone: 356CT10.6.1.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031224EA 587 EUR
USBI031224
Anti-RPS6KA3 Mouse Monoclonal Antibody [clone: 356CT10.6.1.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: RPS6KA3 (ribosomal protein S6 kinase, 90kDa, polypeptide 3)
Clonality: Monoclonal
Clone: 356CT10.6.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR