Imprimer

Anti-RPS6KL1 Mouse Monoclonal Antibody [clone: 273CT4.5.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031229EA 527 EUR
USBI031229
Anti-RPS6KL1 Mouse Monoclonal Antibody [clone: 273CT4.5.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-8000

Type: Primary
Antigen: RPS6KL1 (ribosomal protein S6 kinase-like 1)
Clonality: Monoclonal
Clone: 273CT4.5.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Hamster, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR