Imprimer

Anti-RYK Mouse Monoclonal Antibody [clone: 240CT2.2.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031234EA 495 EUR
USBI031234
Anti-RYK Mouse Monoclonal Antibody [clone: 240CT2.2.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: RYK (receptor-like tyrosine kinase)
Clonality: Monoclonal
Clone: 240CT2.2.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR