Imprimer

Anti-SERPINH1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 134CT3.1.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031246-HRPEA 711 EUR
USBI031246-HRP
Anti-SERPINH1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 134CT3.1.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-200; Immunochemistry: 1:50-100

Type: Primary
Antigen: SERPINH1 (serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1))
Clonality: Monoclonal
Clone: 134CT3.1.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR