Imprimer

Anti-SNX6 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 335CT6.4.6]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031253-FITCEA 711 EUR
USBI031253-FITC
Anti-SNX6 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 335CT6.4.6]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SNX6 (sorting nexin 6)
Clonality: Monoclonal
Clone: 335CT6.4.6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR