Imprimer

Anti-SOX2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 57CT23.3.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031255-APEA 728 EUR
USBI031255-AP
Anti-SOX2 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 57CT23.3.4]
Anticorps
Recommended dilution: ELISA: 1:10000; Western Blot: 1:200-2000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: Monoclonal
Clone: 57CT23.3.4
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR