Imprimer

Anti-SPRED1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 462CT2.5.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031264-FITCEA 711 EUR
USBI031264-FITC
Anti-SPRED1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 462CT2.5.1]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: SPRED1 (sprouty-related, EVH1 domain containing 1)
Clonality: Monoclonal
Clone: 462CT2.5.1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR