Imprimer

Anti-STK39 Mouse Monoclonal Antibody [clone: 430CT4.4.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031267EA 587 EUR
USBI031267
Anti-STK39 Mouse Monoclonal Antibody [clone: 430CT4.4.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-4000

Type: Primary
Antigen: STK39 (serine threonine kinase 39)
Clonality: Monoclonal
Clone: 430CT4.4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR