Imprimer

Anti-SWAP70 Mouse Monoclonal Antibody (Biotin) [clone: 434CT16.5.5]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031271-BIOTINEA 711 EUR
USBI031271-BIOTIN
Anti-SWAP70 Mouse Monoclonal Antibody (Biotin) [clone: 434CT16.5.5]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: SWAP70 (SWAP switching B-cell complex 70kDa subunit)
Clonality: Monoclonal
Clone: 434CT16.5.5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR