Imprimer

Anti-TAOK3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 173CT19.6.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031277-HRPEA 711 EUR
USBI031277-HRP
Anti-TAOK3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 173CT19.6.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:10-50

Type: Primary
Antigen: TAOK3 (TAO kinase 3)
Clonality: Monoclonal
Clone: 173CT19.6.2
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR