Imprimer

Anti-TGFB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 220ct16.4.3.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031281-PEEA 711 EUR
USBI031281-PE
Anti-TGFB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 220ct16.4.3.1]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1000; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50
Do not freeze! Light sensitive

Type: Primary
Antigen: TGFB2 (transforming growth factor, beta 2)
Clonality: Monoclonal
Clone: 220ct16.4.3.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR