Imprimer

Anti-THOC1 Mouse Monoclonal Antibody [clone: 301CT40.5.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031290EA 495 EUR
USBI031290
Anti-THOC1 Mouse Monoclonal Antibody [clone: 301CT40.5.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: THOC1 (THO complex 1)
Clonality: Monoclonal
Clone: 301CT40.5.1
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR