Imprimer

Anti-TIMP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3F10F14]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031291-APCEA 711 EUR
USBI031291-APC
Anti-TIMP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3F10F14]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50

Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: Monoclonal
Clone: 3F10F14
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR