Imprimer

Anti-TLE1 Mouse Monoclonal Antibody (Biotin) [clone: 381CT18.5.1]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031293-BIOTINEA 711 EUR
USBI031293-BIOTIN
Anti-TLE1 Mouse Monoclonal Antibody (Biotin) [clone: 381CT18.5.1]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: TLE1 (transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila))
Clonality: Monoclonal
Clone: 381CT18.5.1
Conjugation: Biotin
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR